The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017344.1 | NP_059040.1 | 10 - 11 | 1499 | 79 |
GenBank mRNA | X53427.1 | - | 10 - 11 | 1499 | 79 |