Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hpcal4
Entrez Gene ID
50872
Gene Name
hippocalcin-like 4
Gene Aliases
NVP-2, Nvjp2
Chromosome Location
Chr.5: 140870128 - 140882242 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017357.1 NP_059053.1 2 - 3 236 75
XM_006238829.2 XP_006238891.1 3 - 4 531 75
GenBank mRNA D13125.1 - 2 - 3 236 75
D14819.1 - - 664 75
FQ212482.1 - 3 - 4 364 75

View Full Product Details