The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017359.2 | NP_059055.2 | 5 - 6 | 567 | 79 |
GenBank mRNA | AY310140.1 | - | 3 - 4 | 331 | 79 |
AY724493.1 | - | 5 - 6 | 746 | 79 | |
BC085744.1 | - | 5 - 6 | 567 | 79 | |
M83677.1 | - | 5 - 6 | 719 | 79 |