The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019146.2 | NP_062019.2 | 5 - 6 | 8717 | 112 |
GenBank mRNA | Y16563.1 | - | 5 - 6 | 8720 | 112 |