The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019166.2 | NP_062039.1 | 3 - 4 | 534 | 61 |
GenBank mRNA | U39549.1 | - | 3 - 4 | 503 | 61 |