The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019187.1 | NP_062060.1 | 4 - 5 | 558 | 104 |
GenBank mRNA | BC081811.1 | - | - | 723 | 104 |
BC098666.1 | - | - | 716 | 104 | |
BC126094.1 | - | - | 730 | 104 | |
FQ216025.1 | - | - | 770 | 104 | |
FQ230994.1 | - | 2 - 3 | 335 | 104 | |
L20427.1 | - | 4 - 5 | 558 | 104 |