The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019190.1 | NP_062063.1 | 1 - 2 | 134 | 59 |
XM_006250478.2 | XP_006250540.1 | 1 - 2 | 1117 | 59 | |
XM_008769849.1 | XP_008768071.1 | 1 - 2 | 1117 | 59 | |
XM_008769850.1 | XP_008768072.1 | 1 - 2 | 883 | 59 | |
GenBank mRNA | AB010920.1 | - | 1 - 2 | 134 | 59 |