The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019255.1 | NP_062128.1 | 1 - 2 | 234 | 73 |
GenBank mRNA | FQ215004.1 | - | 1 - 2 | 331 | 73 |
FQ215168.1 | - | 1 - 2 | 313 | 73 | |
FQ215755.1 | - | 1 - 2 | 313 | 73 | |
FQ216096.1 | - | 1 - 2 | 349 | 73 | |
FQ216734.1 | - | 1 - 2 | 349 | 73 | |
FQ224130.1 | - | 1 - 2 | 405 | 73 | |
FQ224197.1 | - | 1 - 2 | 328 | 73 | |
Y09453.1 | - | 1 - 2 | 234 | 73 |