The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019317.1 | NP_062190.1 | 4 - 5 | 1070 | 70 |
GenBank mRNA | D87840.1 | - | 4 - 5 | 1070 | 70 |