Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Padi1
Entrez Gene ID
54282
Gene Name
peptidyl arginine deiminase, type I
Gene Aliases
Pdi1
Chromosome Location
Chr.5: 159338850 - 159371313 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019332.1 NP_062205.1 6 - 7 780 78
GenBank mRNA AB010998.1 - 6 - 7 780 78

View Full Product Details