Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Haao
Entrez Gene ID
56823
Gene Name
3-hydroxyanthranilate 3,4-dioxygenase
Gene Aliases
-
Chromosome Location
Chr.6: 7045145 - 7059219 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020076.2 NP_064461.1 5 - 6 524 63
XM_006239684.2 XP_006239746.1 5 - 6 1429 63
GenBank mRNA BC085739.1 - 5 - 6 478 63
D28339.1 - 3 - 4 238 63
D44494.1 - 5 - 6 525 63
FQ219702.1 - 5 - 6 538 63

View Full Product Details