Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cnr2
Entrez Gene ID
57302
Gene Name
cannabinoid receptor 2 (macrophage)
Gene Aliases
CB-2, CB2, CB2C, CNR2C
Chromosome Location
Chr.5: 154242214 - 154268126 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA FJ694961.1 - 1 - 2 221 82
JX494784.1 - 1 - 2 221 82

View Full Product Details