The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021763.1 | NP_068531.1 | 6 - 7 | 700 | 74 |
XM_006232539.2 | XP_006232601.1 | 6 - 7 | 961 | 74 | |
XM_006232540.2 | XP_006232602.1 | 7 - 8 | 1103 | 74 | |
XM_006232541.2 | XP_006232603.1 | 8 - 9 | 1170 | 74 | |
XM_006232543.1 | XP_006232605.1 | 6 - 7 | 713 | 74 | |
XM_006232545.2 | XP_006232607.1 | 5 - 6 | 864 | 74 | |
XM_006232546.2 | XP_006232608.1 | 6 - 7 | 1006 | 74 | |
XM_008761110.1 | XP_008759332.1 | 6 - 7 | 717 | 74 | |
XM_008761111.1 | XP_008759333.1 | 6 - 7 | 759 | 74 | |
GenBank mRNA | AY004875.1 | - | 6 - 7 | 700 | 74 |
FQ222383.1 | - | 6 - 7 | 733 | 74 |