The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022191.1 | NP_071527.1 | 1 - 2 | 447 | 95 |
GenBank mRNA | U20105.1 | - | 1 - 2 | 447 | 95 |