The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022250.2 | NP_071586.2 | 3 - 4 | 464 | 58 |
GenBank mRNA | AB034634.1 | - | 3 - 4 | 459 | 58 |
BC097321.1 | - | 3 - 4 | 464 | 58 |