The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022290.1 | NP_071626.1 | 2 - 3 | 398 | 61 |
GenBank mRNA | AB055423.1 | - | 2 - 3 | 187 | 61 |
AF191769.1 | - | 2 - 3 | 398 | 61 |