Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npffr1
Entrez Gene ID
64107
Gene Name
neuropeptide FF receptor 1
Gene Aliases
Gpr147, NPFF1, OT7T022
Chromosome Location
Chr.20: 31214950 - 31248042 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022291.2 NP_071627.2 2 - 3 436 81
XM_006256485.2 XP_006256547.1 2 - 3 582 81
GenBank mRNA AB040103.1 - 2 - 3 428 81
AF268901.1 - 2 - 3 428 81
AF330056.1 - 2 - 3 428 81

View Full Product Details