Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddah1
Entrez Gene ID
64157
Gene Name
dimethylarginine dimethylaminohydrolase 1
Gene Aliases
-
Chromosome Location
Chr.2: 251634368 - 251765026 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022297.2 NP_071633.1 5 - 6 848 69

View Full Product Details