The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022400.1 | NP_071795.1 | 3 - 4 | 311 | 72 |
XM_006229149.1 | XP_006229211.1 | 3 - 4 | 488 | 72 | |
XM_008759413.1 | XP_008757635.1 | 3 - 4 | 1053 | 72 | |
GenBank mRNA | BC061790.1 | - | 3 - 4 | 307 | 72 |
U68417.1 | - | 3 - 4 | 311 | 72 |