Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppt1
Entrez Gene ID
29411
Gene Name
palmitoyl-protein thioesterase 1
Gene Aliases
Ppt
Chromosome Location
Chr.5: 140538260 - 140558163 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022502.2 NP_071947.1 5 - 6 542 60
GenBank mRNA BC126089.1 - 5 - 6 542 60
FQ213474.1 - 5 - 6 554 60
FQ213911.1 - - 554 60
L34262.1 - - 540 60

View Full Product Details