The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022530.1 | NP_071975.1 | 4 - 5 | 366 | 103 |
GenBank mRNA | AF139808.1 | - | 4 - 5 | 366 | 103 |
FQ219962.1 | - | 4 - 5 | 388 | 103 | |
FQ220048.1 | - | 4 - 5 | 387 | 103 | |
FQ220059.1 | - | 4 - 5 | 389 | 103 | |
FQ220802.1 | - | 4 - 5 | 389 | 103 | |
FQ221033.1 | - | 4 - 5 | 388 | 103 | |
FQ222250.1 | - | 4 - 5 | 389 | 103 | |
FQ229694.1 | - | 4 - 5 | 387 | 103 | |
FQ229792.1 | - | 4 - 5 | 388 | 103 | |
FQ229841.1 | - | 4 - 5 | 389 | 103 | |
FQ230178.1 | - | 4 - 5 | 389 | 103 |