The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022614.2 | NP_072136.1 | 1 - 2 | 408 | 87 |
GenBank mRNA | AF140031.1 | - | 1 - 2 | 315 | 87 |
BC089799.1 | - | 1 - 2 | 408 | 87 |