The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022685.2 | NP_073176.2 | 1 - 2 | 188 | 100 |
GenBank mRNA | AF084464.1 | - | 1 - 2 | 62 | 99 |
AY916790.1 | - | 1 - 2 | 128 | 100 | |
BC127495.1 | - | 1 - 2 | 173 | 100 | |
CB706504.1 | - | 1 - 2 | 221 | 100 |