Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sebox
Entrez Gene ID
58922
Gene Name
SEBOX homeobox
Gene Aliases
Og9x
Chromosome Location
Chr.10: 65772443 - 65773314 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023951.1 NP_076441.1 2 - 3 189 59

View Full Product Details