The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023960.2 | NP_076450.1 | 2 - 3 | 484 | 85 |
GenBank mRNA | AB050637.1 | - | 2 - 3 | 466 | 85 |
AY028605.1 | - | 2 - 3 | 484 | 85 | |
BC161829.1 | - | 2 - 3 | 664 | 85 |