The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023965.1 | NP_076455.1 | 3 - 4 | 252 | 77 |
GenBank mRNA | AF298656.3 | - | 3 - 4 | 252 | 77 |
AJ295950.1 | - | 3 - 4 | 252 | 77 | |
FQ217794.1 | - | - | 321 | 77 | |
FQ231233.1 | - | - | 305 | 77 |