Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csf1
Entrez Gene ID
78965
Gene Name
colony stimulating factor 1 (macrophage)
Gene Aliases
-
Chromosome Location
Chr.2: 210522370 - 210550546 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023981.4 NP_076471.3 4 - 5 551 85
XM_008761428.1 XP_008759650.1 4 - 5 422 85
GenBank mRNA AF514355.1 - 4 - 5 391 85
AF514356.1 - 4 - 5 391 85
AF514357.1 - 4 - 5 401 85
AF515736.1 - 4 - 5 489 85
BC074007.1 - 4 - 5 551 85

View Full Product Details