Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pigm
- Entrez Gene ID
-
79112
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis, class M
- Gene Aliases
-
GPI-MT-I
- Chromosome Location
-
Chr.13: 90759260 - 90762957 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_024144.1 |
NP_077058.1 |
1 - 1 |
952 |
121 |
GenBank mRNA |
AB028126.1 |
- |
1 - 1 |
952 |
121 |
- Gene Symbol
- LOC103694910
- Entrez Gene ID
-
103694910
- Gene Name
- GPI mannosyltransferase 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Un|NW_007906966: 760 - 7190 on Build Rnor_6.0
View Full Product Details