The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024400.2 | NP_077376.2 | 1 - 2 | 1107 | 107 |
GenBank mRNA | AF149118.1 | - | 1 - 2 | 1120 | 107 |
BC080237.1 | - | 1 - 2 | 1107 | 107 |