Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sost
Entrez Gene ID
80722
Gene Name
sclerostin
Gene Aliases
-
Chromosome Location
Chr.10: 89897087 - 89900131 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030584.1 NP_085073.1 1 - 2 257 81
GenBank mRNA AF326741.1 - 1 - 2 257 81

View Full Product Details