The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030828.1 | NP_110455.1 | 1 - 2 | 387 | 116 |
GenBank mRNA | BC061572.1 | - | 1 - 2 | 385 | 116 |
L02896.1 | - | 1 - 2 | 387 | 116 | |
L34067.1 | - | 1 - 2 | 220 | 116 |