The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030829.1 | NP_110456.1 | 8 - 9 | 919 | 83 |
XM_006231686.1 | XP_006231748.1 | 8 - 9 | 972 | 83 | |
GenBank mRNA | BC081792.1 | - | 8 - 9 | 972 | 83 |
U34841.1 | - | 8 - 9 | 919 | 83 |