The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030853.1 | NP_110480.1 | 8 - 9 | 738 | 68 |
XM_008759919.1 | XP_008758141.1 | 7 - 8 | 1162 | 68 | |
GenBank mRNA | AJ001184.1 | - | 8 - 9 | 738 | 68 |
BC088130.1 | - | 7 - 8 | 766 | 68 | |
FQ233731.1 | - | 7 - 8 | 725 | 68 |