Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrn3
Entrez Gene ID
81514
Gene Name
leucine rich repeat neuronal 3
Gene Aliases
Nlrr3
Chromosome Location
Chr.6: 61374328 - 61405195 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030856.1 NP_110483.1 1 - 2 334 70
GenBank mRNA AF291437.1 - 1 - 2 334 70

View Full Product Details