The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031034.2 | NP_112296.1 | 2 - 3 | 548 | 77 |
GenBank mRNA | D85760.1 | - | 2 - 3 | 582 | 77 |
DQ120479.1 | - | 2 - 3 | 517 | 77 | |
DQ120480.1 | - | 2 - 3 | 517 | 77 |