The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031085.2 | NP_112347.1 | 7 - 8 | 1144 | 75 |
XM_008764720.1 | XP_008762942.1 | 7 - 8 | 1026 | 75 | |
GenBank mRNA | BC061763.1 | - | 8 - 9 | 1475 | 75 |
BC081782.1 | - | 7 - 8 | 1144 | 75 | |
X68400.1 | - | 7 - 8 | 985 | 75 |