The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031131.1 | NP_112393.1 | 2 - 3 | 742 | 95 |
GenBank mRNA | AF153012.2 | - | 2 - 3 | 742 | 95 |