The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031590.1 | NP_113778.1 | 2 - 3 | 322 | 124 |
GenBank mRNA | AF259981.1 | - | 2 - 3 | 322 | 124 |