The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031692.2 | NP_113880.2 | 3 - 4 | 281 | 113 |
GenBank mRNA | AF033027.1 | - | 3 - 4 | 281 | 113 |
AY881621.1 | - | 3 - 4 | 281 | 113 |