Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St3gal3
Entrez Gene ID
64445
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Gene Aliases
Siat6
Chromosome Location
Chr.5: 136765309 - 136965642 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031697.1 NP_113885.1 9 - 10 822 66
XM_006238715.2 XP_006238777.1 9 - 10 1259 66
XM_006238716.2 XP_006238778.1 8 - 9 1225 66
XM_008764066.1 XP_008762288.1 9 - 10 3749 66
XM_008764067.1 XP_008762289.1 9 - 10 842 66
GenBank mRNA M97754.1 - 9 - 10 822 66

View Full Product Details