The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031730.2 | NP_113918.2 | 4 - 5 | 2020 | 127 |
XM_008762830.1 | XP_008761052.1 | 4 - 5 | 544 | 127 | |
GenBank mRNA | CO393575.1 | - | 4 - 5 | 581 | 127 |
M59980.1 | - | 4 - 5 | 1697 | 125 | |
S64320.1 | - | 4 - 5 | 2018 | 127 |