The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031771.2 | NP_113959.1 | 1 - 1 | 1625 | 68 |
GenBank mRNA | AF022743.1 | - | 1 - 1 | 1550 | 68 |
BC070903.1 | - | 1 - 1 | 1625 | 68 |