The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031773.1 | NP_113961.1 | 3 - 4 | 556 | 95 |
GenBank mRNA | AF025424.1 | - | 3 - 4 | 556 | 95 |