Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aldh3a1
Entrez Gene ID
25375
Gene Name
aldehyde dehydrogenase 3 family, member A1
Gene Aliases
AHDC, Ahd-c, Aldh, Aldh3
Chromosome Location
Chr.10: 47490168 - 47499855 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031972.1 NP_114178.1 1 - 2 173 100
GenBank mRNA BC070924.1 - 1 - 2 206 100
J03637.1 - 1 - 2 173 100

View Full Product Details