Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clta
Entrez Gene ID
83800
Gene Name
clathrin, light chain A
Gene Aliases
LCA1, LCA2, LCA3
Chromosome Location
Chr.5: 59490363 - 59509139 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031974.1 NP_114180.1 4 - 5 603 68
XM_006238104.1 XP_006238166.1 4 - 5 1376 68
GenBank mRNA FQ213034.1 - 4 - 5 611 68
M15882.1 - 4 - 5 603 68
M19260.1 - 4 - 5 603 68

View Full Product Details