The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033098.2 | NP_149089.1 | 3 - 4 | 491 | 58 |
GenBank mRNA | AJ400732.1 | - | 3 - 4 | 480 | 58 |
BC062007.1 | - | 3 - 4 | 482 | 58 | |
FM054278.1 | - | 3 - 4 | 482 | 58 | |
KC222917.1 | - | 3 - 4 | 468 | 58 | |
KC222918.1 | - | 3 - 4 | 468 | 58 | |
KC222919.1 | - | 3 - 4 | 468 | 58 | |
KC222920.1 | - | 3 - 4 | 468 | 58 |