The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052980.2 | NP_443212.1 | 8 - 9 | 1395 | 59 |
GenBank mRNA | AF151377.1 | - | 8 - 9 | 1368 | 59 |
BC091138.1 | - | 7 - 8 | 1489 | 59 | |
BC107449.1 | - | 6 - 7 | 1270 | 59 |