Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ssbp3
Entrez Gene ID
84354
Gene Name
single stranded DNA binding protein 3
Gene Aliases
Ssdp, Ssdp3
Chromosome Location
Chr.5: 126508099 - 126649830 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053358.1 NP_445810.1 4 - 5 625 93
XM_006238532.2 XP_006238594.1 4 - 5 660 93
XM_006238533.2 XP_006238595.1 4 - 5 659 93
XM_006238534.2 XP_006238596.1 4 - 5 659 93
XM_006238535.2 XP_006238597.1 4 - 5 657 93
XM_006238536.2 XP_006238598.1 4 - 5 658 93
XM_006238537.2 XP_006238599.1 4 - 5 656 93
XM_006238538.2 XP_006238600.1 4 - 5 656 93
XM_008763902.1 XP_008762124.1 4 - 5 2313 93
XM_008763903.1 XP_008762125.1 5 - 6 1867 93
XM_008763904.1 XP_008762126.1 4 - 5 2310 93
XM_008763905.1 XP_008762127.1 4 - 5 2310 93
XM_008763906.1 XP_008762128.1 4 - 5 2307 93
XM_008763907.1 XP_008762129.1 4 - 5 2309 93
XM_008763908.1 XP_008762130.1 4 - 5 2307 93
XM_008763909.1 XP_008762131.1 4 - 5 2304 93
GenBank mRNA AF121893.1 - 4 - 5 625 93
AY724490.1 - 4 - 5 325 93
BC107666.1 - 4 - 5 609 93

View Full Product Details