The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053375.1 | NP_445827.1 | 2 - 3 | 889 | 121 |
GenBank mRNA | AF247450.1 | - | 2 - 3 | 889 | 121 |