The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053402.2 | NP_445854.1 | 2 - 3 | 318 | 69 |
GenBank mRNA | AF188608.1 | - | 2 - 3 | 319 | 69 |
BC098752.1 | - | 2 - 3 | 339 | 69 |